Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RAMP2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6427
This item is not returnable.
View return policy
Description
RAMP2 Polyclonal specifically detects RAMP2 in Human samples. It is validated for Western Blot.Specifications
RAMP2 | |
Polyclonal | |
Unconjugated | |
O60895 | |
RAMP2 | |
E. coli-derived recombinant human RAMP2 Gln43-Val145 Accession # O60895 | |
100 μg | |
Primary | |
Detects human RAMP2 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse RAMP2 is observed and less than 1% cross-reactivity with recombinant human (rh) RAMP1 and rhRAMP3. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
calcitonin receptor-like receptor activity modifying protein 2, Calcitonin-receptor-like receptor activity-modifying protein 2, CRLR activity-modifying protein 2, receptor (calcitonin) activity modifying protein 2, receptor (G protein-coupled) activity modifying protein 2, receptor activity modifying protein 2, receptor activity-modifying protein 2, receptor-activity-modifying protein 2 | |
Sheep | |
Affinity Purified | |
RUO | |
10266 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction