Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RANK/TNFRSF11A Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $2020.00
Specifications
| Antigen | RANK/TNFRSF11A |
|---|---|
| Clone | 80707 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB6831SP
![]() |
R&D Systems
MAB6831SP |
25 μg |
Each for $200.50
|
|
|||||
MAB6831100
![]() |
R&D Systems
MAB6831100 |
100 μg |
Each for $549.50
|
|
|||||
MAB6831500
![]() |
R&D Systems
MAB6831500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| RANK/TNFRSF11A | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q9Y6Q6 | |
| 8792 | |
| Mouse myeloma cell line NS0-derived recombinant human RANK Leu28-Gly213 Accession # Q9Y6Q6 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant human RANK in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, recombinant mouse (rm) BAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR4, rhDR5, rhEDAR, rmEDAR, rmFas, rhGITR, rhHVEM, rhLymphotoxin beta R, rhNGF R, rhOPG, rmRANK, rhRELT, rhTNF RI, rmTNF RI, rhTNF RII, rhTRAIL R2, rhTRAIL R3, rhTRAIL R4, or rhTROY is observed. |
| 80707 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFR, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TNFRSF11A, TRANCE R, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| TNFRSF11A | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title