Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RANK/TNFRSF11A Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF683
This item is not returnable.
View return policy
Description
RANK/TNFRSF11A Polyclonal specifically detects RANK/TNFRSF11A in Human samples. It is validated for Western Blot, Agonist Activity.Specifications
RANK/TNFRSF11A | |
Polyclonal | |
Unconjugated | |
Q9Y6Q6 | |
TNFRSF11A | |
Mouse myeloma cell line NS0-derived recombinant human RANK Gln29-Gly213 Accession # Q9Y6Q6 | |
100 μg | |
Agonist Activity | |
8792 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL, Agonist Activity 4-8 ug/mL | |
CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFR, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TNFRSF11A, TRANCE R, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Detects human RANK in direct ELISAs and Western blots. In these formats, approximately 30% cross-reactivity with recombinant mouse RANK is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction