Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RAR beta/NR1B2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems PPH433800
This item is not returnable.
View return policy
Description
RAR beta/NR1B2 Monoclonal specifically detects RAR beta/NR1B2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Direct ELISA.Specifications
RAR beta/NR1B2 | |
Monoclonal | |
Unconjugated | |
A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
RARB | |
Recombinant human RAR beta/NR1B2 aa 2-79 | |
100 μg | |
Primary | |
This antibody specifically recognizes human RAR beta and does not cross-react with human RAR alpha or RAR gamma . Not yet tested in other species. | |
This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunoprecipitation, ELISA | |
H4338 | |
Western Blot, Immunoprecipitation, Direct ELISA | |
beta polypeptide, HAP, HBV-activated protein, NR1B2, RARB, retinoic acid receptor, beta, RRB2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5915 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction