Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RASGRP4 Full-length ORF (NP_733749.1, 1 a.a. - 673 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89959461
Click to view available options
Quantity:
2 μg

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the Ras guanyl nucleotide-releasing protein (RasGRP) family of Ras guanine nucleotide exchange factors. It contains a Ras exchange motif, a diacylglycerol-binding domain, and two calcium-binding EF hands. This protein was shown to activate H-Ras in a cation-dependent manner in vitro. Expression of this protein in myeloid cell lines was found to be correlated with elevated level of activated RAS protein, and the RAS activation can be greatly enhanced by phorbol ester treatment, which suggested a role of this protein in diacylglycerol regulated cell signaling pathways. Studies of a mast cell leukemia cell line expressing substantial amounts of abnormal transcripts of this gene indicated that this gene may play an important role in the final stages of mast cell development. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MNRKDSKRKSHQECTGKIGGRGRPRQVRRHKTCPSPREISKVMASMNLGLLSEGGCSEDELLEKCIQSFDSAGSLCHEDHMLNMVLAMHSWVLPSADLAARLLTSYQKATGDTQELRRLQICHLVRYWLMRHPEVMHQDPQLEEVIGRFWATVAREGNSAQRRLGDSSDLLSPGGPGPPLPMSSPGLGKKRKVSLLFDHLETGELAQHLTYLEFRSFQAITPQDLRSYVLQGSVRGCPALEGSVGLSNSVSRWVQVMVLSRPGPLQRAQVLDKFIHVAQRLHQLQNFNTLMAVTGGLCHSAISRLKDSHAHLSPDSTKALLELTELLASHNNYARYRRTWAGCAGFRLPVLGVHLKDLVSLHEAQPDRLPDGRLHLPKLNNLYLRLQELVALQGQHPPCSANEDLLHLLTLSLDLFYTEDEIYELSYAREPRCPKSLPPSPFNAPLVVEWAPGVTPKPDRVTLGRHVEQLVESVFKNYDPEGRGTISQEDFERLSGNFPFACHGLHPPPRQGRGSFSREELTGYLLRASAICSKLGLAFLHTFHEVTFRKPTFCDSCSGFLWGVTKQGYRCRECGLCCHKHCRDQVKVECKKRPGAKGDAGPPGAPVPSTPAPHASCGSEENHSYTLSLEPETGCQLRHAWTQTESPHPSWETDTVPCPVMDPPSTASSKLDS

Specifications

Accession Number NP_733749.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 115727
Molecular Weight (g/mol) 101.3kDa
Name RASGRP4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MNRKDSKRKSHQECTGKIGGRGRPRQVRRHKTCPSPREISKVMASMNLGLLSEGGCSEDELLEKCIQSFDSAGSLCHEDHMLNMVLAMHSWVLPSADLAARLLTSYQKATGDTQELRRLQICHLVRYWLMRHPEVMHQDPQLEEVIGRFWATVAREGNSAQRRLGDSSDLLSPGGPGPPLPMSSPGLGKKRKVSLLFDHLETGELAQHLTYLEFRSFQAITPQDLRSYVLQGSVRGCPALEGSVGLSNSVSRWVQVMVLSRPGPLQRAQVLDKFIHVAQRLHQLQNFNTLMAVTGGLCHSAISRLKDSHAHLSPDSTKALLELTELLASHNNYARYRRTWAGCAGFRLPVLGVHLKDLVSLHEAQPDRLPDGRLHLPKLNNLYLRLQELVALQGQHPPCSANEDLLHLLTLSLDLFYTEDEIYELSYAREPRCPKSLPPSPFNAPLVVEWAPGVTPKPDRVTLGRHVEQLVESVFKNYDPEGRGTISQEDFERLSGNFPFACHGLHPPPRQGRGSFSREELTGYLLRASAICSKLGLAFLHTFHEVTFRKPTFCDSCSGFLWGVTKQGYRCRECGLCCHKHCRDQVKVECKKRPGAKGDAGPPGAPVPSTPAPHASCGSEENHSYTLSLEPETGCQLRHAWTQTESPHPSWETDTVPCPVMDPPSTASSKLDS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name RASGRP4
Gene Symbol RASGRP4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.