Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat Brevican Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4009
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4009 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4009 Supplier R&D Systems Supplier No. AF4009
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 4 publications

Brevican Polyclonal specifically detects Brevican in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen Brevican
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. AAH27971
Gene Alias ALPBRE, BCAN, BEHAB, BEHABbrevican core protein, Brain-enriched hyaluronan-binding protein, brevican, Chondroitin sulfate proteoglycan 7, chondroitin sulfate proteoglycan BEHAB, chondroitin sulfate proteoglycan BEHAB/brevican, CSPG7, CSPG7brevican proteoglycan, MGC13038
Gene Symbols BCAN
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Brevican Asp23-Pro911 Accession # AAH27971
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63827
Test Specificity Detects human Brevican in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Brevican is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.