Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat CCR4 PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1567C100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1567C100 100 Tests
FAB1567C025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1567C100 Supplier R&D Systems Supplier No. FAB1567C100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

CCR4 Monoclonal specifically detects CCR4 in Human, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR4
Applications Flow Cytometry
Classification Monoclonal
Clone 205410
Conjugate PerCP
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. P51679
Gene Alias C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5
Gene Symbols CCR4
Host Species Mouse
Immunogen Human CCR4 transfectants Met1-Leu360 Accession # P51679
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1233
Test Specificity Stains human CCR4-transfected cell lines but not the corresponding parent cell lines. Also detects human CCR4 on human PBLs and platelets and rat CCR4 on rat splenocytes.
Target Species Human, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.