Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat/Chicken Laminin S Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB2066
This item is not returnable.
View return policy
Description
Laminin S/Laminin beta 2 Monoclonal specifically detects Laminin S/Laminin beta 2 in Human, Rat, Chicken samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.Specifications
Laminin S/Laminin beta 2 | |
Monoclonal | |
LYOPH | |
Western Blot, Immunoprecipitation, Immunocytochemistry 8-25 ug/mL | |
LAMB2, Laminin B1s chain, Laminin beta 2, laminin S, laminin subunit beta-2, laminin, beta 2 (laminin S), Laminin-11 subunit beta, Laminin-14 subunit beta, Laminin-15 subunit beta, Laminin-3 subunit beta, Laminin-4 subunit beta, Laminin-7 subunit beta, LAMSLaminin-9 subunit beta, S-LAM beta, S-laminin subunit beta | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3913 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunoprecipitation, Immunocytochemistry | |
C4 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LAMB2 | |
Bovine anterior lens capsule | |
100 μg | |
Primary | |
Detects human, rat, and chicken Laminin S in Western blots. | |
Human, Rat, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction