Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat Cortactin Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6096
This item is not returnable.
View return policy
Description
Cortactin Polyclonal specifically detects Cortactin in Human, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
Cortactin | |
Polyclonal | |
Unconjugated | |
Q14247 | |
CTTN | |
E. coli-derived recombinant human Cortactin Met1-Gly85 Accession # Q14247 | |
100 μg | |
Primary | |
Detects human and rat Cortactin in Western blots. | |
Human, Rat | |
IgG |
Western Blot, Immunoassay | |
LYOPH | |
Western Blot 1 ug/mL, Knockout Validated | |
1110020L01Rik, Amplaxin, cortactin, CTTN, EMS1, ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 srcsubstrate), EMS1amplaxin, FLJ34459, Oncogene EMS1, src substrate cortactin | |
Goat | |
Affinity Purified | |
RUO | |
2017 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction