Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat Cystatin B Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1408SP
This item is not returnable.
View return policy
Description
Cystatin B/Stefin B Polyclonal specifically detects Cystatin B/Stefin B in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Knockout Validated.Specifications
Cystatin B/Stefin B | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, CSTB, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B | |
Goat | |
Affinity Purified | |
RUO | |
1476 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoassay | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Knockout Validated | |
P04080 | |
CSTB | |
E. coli-derived recombinant human Cystatin B Met2-Phe98 Accession # P04080 | |
25 μg | |
Primary | |
Detects human and rat Cystatin B in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human (rh) Cystatin A, recombinant mouse Cystatin B, rhCystatin C, rhCystatin D, rhCystatin E/M, rhCystatin S, rhCystatin SA, and rhCystatin SN is observed. | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction