Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat DOCK1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF7070SP
This item is not returnable.
View return policy
Description
DOCK1 Polyclonal specifically detects DOCK1 in Human, Rat samples. It is validated for Western Blot.Specifications
DOCK1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
180 kDa protein downstream of CRK, ced5, dedicator of cytokinesis 1, dedicator of cyto-kinesis 1, DOCK180, DOCK180dedicator of cytokinesis protein 1, DOwnstream of CrK | |
Goat | |
Affinity Purified | |
RUO | |
1793 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q14185 | |
DOCK1 | |
E. coli-derived recombinant human DOCK1 Gly422-Ser666 Accession # Q14185 | |
25 μg | |
Primary | |
Detects recombinant human DOCK1 in direct ELISAs. Detects human and rat DOCK1 in Western blots. | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction