Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat EphA5 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB541G100UG
This item is not returnable.
View return policy
Description
EphA5 Monoclonal specifically detects EphA5 in Human, Rat samples. It is validated for Flow Cytometry.Specifications
EphA5 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P54757 | |
EPHA5 | |
Mouse myeloma cell line NS0-derived recombinant rat EphA5 Ser58-Gln573 (Asp170Glu) Accession # P54757 | |
RUO | |
2044 | |
Human, Rat | |
IgG1 |
Flow Cytometry | |
86731 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Bsk, Cek7, EC 2.7.10, EC 2.7.10.1, Ehk1, EHK-1, EHK1Hek7, EK7, EPH homology kinase 1, Eph homology kinase-1, EPH receptor A5, EphA5, EPH-like kinase 7, ephrin type-A receptor 5, hEK7, receptor protein-tyrosine kinase HEK7, Rek7, TYRO4HEK7CEK7, tyrosine-protein kinase receptor EHK-1 | |
Mouse | |
100 μg | |
Primary | |
Detects rat EphA5 in direct ELISAs and Western blots. In direct ELISAs, 5-15% cross reactivity with recombinant mouse (rm) EphA4 and rmEpha8 is observed and no cross-reactivity with recombinant human (rh) EphA1, rmEphA2, -A3, -A6, -A7, recombinant rat EphB1, -B2, or rmEphB3 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction