Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat EphA5 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB541S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB541S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB541S100 Supplier R&D Systems Supplier No. FAB541S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

EphA5 Monoclonal specifically detects EphA5 in Human, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen EphA5
Applications Flow Cytometry
Classification Monoclonal
Clone 86731
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P54757
Gene Alias Bsk, Cek7, EC 2.7.10, EC 2.7.10.1, Ehk1, EHK-1, EHK1Hek7, EK7, EPH homology kinase 1, Eph homology kinase-1, EPH receptor A5, EphA5, EPH-like kinase 7, ephrin type-A receptor 5, hEK7, receptor protein-tyrosine kinase HEK7, Rek7, TYRO4HEK7CEK7, tyrosine-protein kinase receptor EHK-1
Gene Symbols EPHA5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat EphA5 Ser58-Gln573 (Asp170Glu) Accession # P54757
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2044
Test Specificity Detects rat EphA5 in direct ELISAs and Western blots. In direct ELISAs, 5-15% cross reactivity with recombinant mouse (rm) EphA4 and rmEpha8 is observed and no cross-reactivity with recombinant human (rh) EphA1, rmEphA2, -A3, -A6, -A7, recombinant rat EphB1, -B2, or rmEphB3 is observed.
Target Species Human, Rat
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.