Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat EphA5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF541SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF541SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF541SP Supplier R&D Systems Supplier No. AF541SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

EphA5 Polyclonal specifically detects EphA5 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen EphA5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P54757
Gene Alias Bsk, Cek7, EC 2.7.10, EC 2.7.10.1, Ehk1, EHK-1, EHK1Hek7, EK7, EPH homology kinase 1, Eph homology kinase-1, EPH receptor A5, EphA5, EPH-like kinase 7, ephrin type-A receptor 5, hEK7, receptor protein-tyrosine kinase HEK7, Rek7, TYRO4HEK7CEK7, tyrosine-protein kinase receptor EHK-1
Gene Symbols EPHA5
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat EphA5 Met1-Gln573 Accession # P54757
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2044
Test Specificity Detects rat EphA5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse (rm) EphA4, rmEphA3, rmEphA6, rmEphA7, rmEphA8, and recombinant rat EphB1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.