Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat FABP1/L-FABP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | FABP1/L-FABP |
---|---|
Clone | 965608 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB29643SP
![]() |
R&D Systems
MAB29643SP |
25 μg |
Each for $200.50
|
|
|||||
MAB29643100
![]() |
R&D Systems
MAB29643100 |
100 μg |
Each for $502.50
|
|
|||||
Description
FABP1/L-FABP Monoclonal specifically detects FABP1/L-FABP in Human, Rat samples. It is validated for Western Blot, Simple Western, ELISA.Specifications
FABP1/L-FABP | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
P07148 | |
2168 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
965608 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, LFABP, L-FABP, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein | |
E. coli-derived recombinant human FABP1/L-FABP Met1-Ile127 Accession # P07148 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FABP1/L-FABP in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title