Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat GOLGB1/Giantin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8159
This item is not returnable.
View return policy
Description
GOLGB1/Giantin Polyclonal specifically detects GOLGB1/Giantin in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
GOLGB1/Giantin | |
Polyclonal | |
Unconjugated | |
Q14789 | |
GOLGB1 | |
E. coli-derived recombinant human GOLGB1/Giantin Ser1596-Glu1702 Accession # Q14789 | |
100 μg | |
Primary | |
Detects human GOLGB1/Giantin in direct ELISAs. Detects human and rat GOLGB1/Giantin in Western blots. | |
Human, Rat | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
372 kDa Golgi complex-associated protein, GCP, GCP372, GCP372GIANTIN, giantin, golgi autoantigen, golgin subfamily b, macrogolgin (with transmembrane signal)1,macrogolgin, golgi integral membrane protein 1, golgin B1, golgin B1, golgi integral membrane protein, Golgin subfamily B member 1, GOLIM1, Macrogolgin | |
Sheep | |
Affinity Purified | |
RUO | |
2804 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction