Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat GR/NR3C1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10144SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10144SP 25 μg
MAB10144100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10144SP Supplier R&D Systems Supplier No. MAB10144SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GR/NR3C1 Monoclonal specifically detects GR/NR3C1 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen GR/NR3C1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1006903
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P04150
Gene Alias GCR, glucocorticoid receptor, GRGCCR, GRLNuclear receptor subfamily 3 group C member 1, NR3C1, nuclear receptor subfamily 3, group C, member 1, nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor)
Host Species Mouse
Immunogen E. coli-derived recombinant human GR/NR3C1, Val271-Lys777, Accession No. P04150
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 2908.0
Test Specificity Detects human GR/NR3C1 in direct ELISAs. Detects human and rat GR/NR3C1 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.