Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat HIF-2 alpha/EPAS1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF2886SP
This item is not returnable.
View return policy
Description
HIF-2 alpha/EPAS1 Polyclonal specifically detects HIF-2 alpha/EPAS1 in Human, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation (ChIP).Specifications
HIF-2 alpha/EPAS1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Basic-helix-loop-helix-PAS protein MOP2, BHLHE73, Class E basic helix-loop-helix protein 73, ECYT4, endothelial PAS domain protein 1, endothelial PAS domain-containing protein 1, EPAS1, EPAS-1, HIF 2A, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF2 alpha, HIF-2 alpha, HIF2A, hif2a angiogenesis, HIF-2-alpha, HIF2-alpha, HLF, hypoxia-inducible factor 2 alpha, Hypoxia-inducible factor 2-alpha, Member of PAS protein 2, MOP2, PAS domain-containing protein 2, PASD2 | |
Goat | |
Affinity Purified | |
RUO | |
2034 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
ChIP Assay | |
LYOPH | |
Western Blot 1 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells | |
Q99814 | |
EPAS1 | |
E. coli-derived recombinant human HIF-2 alpha/EPAS1 Ser543-Thr870 Accession # Q99814 | |
25 μg | |
Primary | |
Detects human and rat HIF-2 alpha/EPAS1. | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction