Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat IkB-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3425SP
This item is not returnable.
View return policy
Description
IkB-beta Monoclonal specifically detects IkB-beta in Human, Rat samples. It is validated for Western Blot.Specifications
IkB-beta | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q15653 | |
NFKBIB | |
E. coli-derived recombinant human IkB-beta Ala2-Val356 Accession # Q15653 | |
25 μg | |
Primary | |
Detects human and rat IkB-beta in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) IkB-alpha or rhIkB-epsilon is observed. | |
Human, Rat | |
Purified |
Western Blot | |
347428 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkBbeta, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, NFKBIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4793 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction