Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat IkB-beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3425
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3425 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3425 Supplier R&D Systems Supplier No. MAB3425
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IkB-beta Monoclonal specifically detects IkB-beta in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen IkB-beta
Applications Western Blot
Classification Monoclonal
Clone 347428
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q15653
Gene Alias beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkBbeta, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, NFKBIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta
Gene Symbols NFKBIB
Host Species Mouse
Immunogen E. coli-derived recombinant human IkB-beta Ala2-Val356 Accession # Q15653
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4793
Test Specificity Detects human and rat IkB-beta in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) IkB-alpha or rhIkB-epsilon is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.