Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat IL-32 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6769100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6769100 100 μg
MAB6769SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6769100 Supplier R&D Systems Supplier No. MAB6769100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IL-32 Monoclonal specifically detects IL-32 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen IL-32
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2525B
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. NP_001012649
Gene Alias IL32, IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4, NK4IL-32delta, TAIF, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor
Host Species Rabbit
Immunogen E. coli-derived human IL-32 beta protein, Met1-Lys188, Accession # NP_001012649
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9235
Test Specificity Detects human IL-32 beta and other IL-32 isoforms in direct ELISAs. In direct ELISAs, 95% cross-reactivity with recombinant human (rh) IL-32 alpha and 45% cross-reactivity with rhIL-32 gamma is detected.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.