Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat LRP-4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5948
This item is not returnable.
View return policy
Description
LRP-4 Polyclonal specifically detects LRP-4 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LRP-4 | |
Polyclonal | |
Unconjugated | |
O75096 | |
LRP4 | |
E. coli-derived recombinant human LRP-4 Gly18-Ser344 Accession # O75096 | |
100 μg | |
Primary | |
Detects human and rat LRP-4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) LRP-1, rhLRP-1B, rhLRP-5, rhLRP-6, and rhLDL R is observed. | |
Human, Rat | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
CLSS, KIAA0816, low density lipoprotein receptor-related protein 4, low-density lipoprotein receptor-related protein 4, LRP10, LRP4, LRP-4, MEGF7LRP10, Multiple epidermal growth factor-like domains 7 | |
Sheep | |
Affinity Purified | |
RUO | |
4038 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction