Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat M-Ras/R-Ras3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3606SP
This item is not returnable.
View return policy
Description
M-Ras/R-Ras3 Polyclonal specifically detects M-Ras/R-Ras3 in Human, Rat samples. It is validated for Western Blot.Specifications
M-Ras/R-Ras3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ42964, MRas, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs | |
Goat | |
Affinity Purified | |
RUO | |
22808 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
O14807 | |
MRAS | |
E. coli-derived recombinant human M-Ras Met1-Leu208 (Lys181Glu) Accession # O14807 | |
25 μg | |
Primary | |
Detects human and rat M-Ras/R-Ras3 in Western blots. Does not detect recombinant human R-Ras or TC21 (R-Ras2). | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction