Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat M-Ras/R-Ras3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3606
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3606 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3606 Supplier R&D Systems Supplier No. AF3606
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

M-Ras/R-Ras3 Polyclonal specifically detects M-Ras/R-Ras3 in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen M-Ras/R-Ras3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Gene Accession No. O14807
Gene Alias FLJ42964, MRas, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs
Gene Symbols MRAS
Host Species Goat
Immunogen E. coli-derived recombinant human M-Ras Met1-Leu208 (Lys181Glu) Accession # O14807
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22808
Test Specificity Detects human and rat M-Ras/R-Ras3 in Western blots. Does not detect recombinant human R-Ras or TC21 (R-Ras2).
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.