Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat MafB Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3810SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3810SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3810SP Supplier R&D Systems Supplier No. MAB3810SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MafB Monoclonal specifically detects MafB in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen MafB
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 372239
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y5Q3
Gene Alias Kreisler (mouse) maf-related leucine zipper homolog, Kreisler maf-related leucine zipper homolog, KRML, KRMLMAFB/Kreisler basic region/leucine zipper transcription factor, Maf-B, MGC43127, transcription factor MafB, V-maf musculoaponeurotic fibrosarcoma oncogene homolog B, v-maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian)
Gene Symbols MAFB
Host Species Mouse
Immunogen E. coli-derived recombinant human MafB Met1-Leu323 Accession # Q9Y5Q3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9935
Test Specificity Detects human and rat MafB at in Western blots. No cross-reactivity with recombinant human (rh) MafF, rhMafG, or rhMafK is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.