Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat MDGA2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5184RSP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5184R-SP 25 μg
MAB5184R-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5184R-SP Supplier R&D Systems Supplier No. MAB5184RSP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MDGA2 Monoclonal specifically detects MDGA2 in Human, Rat samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen MDGA2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 803732R
Conjugate Unconjugated
Dilution Flow Cytometry 0.25μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. Q7Z553
Gene Alias MAM domain containing glycosylphosphatidylinositol anchor 2, MAMDC1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MDGA2 Gln21-Asp934 Accession # Q7Z553
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 161357
Test Specificity Detects human MDGA2 in direct ELISAs. Detects rat MDGA2 in Flow Cytometry.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.