Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat Myoglobin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $410.00
Specifications
| Antigen | Myoglobin |
|---|---|
| Clone | 974102 |
| Dilution | Western Blot 0.05 ug/mL, Immunohistochemistry 1-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9720SP
![]() |
R&D Systems
MAB9720SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9720100
![]() |
R&D Systems
MAB9720100 |
100 μg |
Each for $410.00
|
|
|||||
Description
Myoglobin Monoclonal specifically detects Myoglobin in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
| Myoglobin | |
| Western Blot 0.05 ug/mL, Immunohistochemistry 1-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human, Rat | |
| P02144 | |
| 4151 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 974102 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MB, MGC13548, myoglobin, PVALB | |
| Purified human cardiac Myoglobin antigen from human heart Accession # P02144 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Myoglobin in direct ELISAs. Detects human and rat Myoglobin in Western blots. In Western blots, no detection of mouse myglobin was observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title