Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat NCAM-1/CD56 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | NCAM-1/CD56 |
---|---|
Clone | 2524F |
Dilution | Western Blot 1 μg/mL, Immunocytochemistry 3-25 μg/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB24085SP
![]() |
R&D Systems
MAB24085SP |
25 μg |
Each for $200.50
|
|
|||||
MAB24085100
![]() |
R&D Systems
MAB24085100 |
100 μg |
Each for $479.50
|
|
|||||
Description
NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
NCAM-1/CD56 | |
Western Blot 1 μg/mL, Immunocytochemistry 3-25 μg/mL | |
Monoclonal | |
Supernatant | |
Human, Rat | |
NP_001070150 | |
4684 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2524F | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD56, CD56/ NCAM-1, CD56 antigen, MSK39, NCAM1, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
Mouse myeloma cell line NS0-derived recombinant human NCAM-1/CD56, Leu20-Pro603 and Glu636-Asn741, Accession # NP_001070150 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human NCAM-1/CD56 in direct ELISAs. Detects human and rat NCAM-1/CD56 in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title