Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat Neurogenin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
$214.00 - $561.00
Specifications
Antigen | Neurogenin-2 |
---|---|
Clone | 7G4 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Description
Neurogenin-2 Monoclonal specifically detects Neurogenin-2 in Human, Rat samples. It is validated for Immunohistochemistry.Specifications
Neurogenin-2 | |
LYOPH | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human, Rat | |
Atoh4, BHLHA8, bHLHa8MGC46562, Class A basic helix-loop-helix protein 8, Math4A, NEUROG2, neurogenin 2, neurogenin-2, Neurogenin2, NGN2, NGN-2, Protein atonal homolog 4 | |
NEUROG2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
7G4 | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
63973 | |
E. coli-derived recombinant rat Neurogenin-2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detect human and rat Neurogenin-2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title