Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat Neuroligin 1/NLGN1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4340
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4340 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4340 Supplier R&D Systems Supplier No. AF4340
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Neuroligin 1/NLGN1 Polyclonal specifically detects Neuroligin 1/NLGN1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Neuroligin 1/NLGN1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q62765
Gene Alias EC 3.1.1, EC 3.1.1.1, KIAA1070neuroligin-1, MGC45115, neuroligin 1, NL1, Nlg, NLGN1
Gene Symbols NLGN1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Neuroligin 1/NLGN1 Gln46-Leu696 Accession # Q62765
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22871
Test Specificity Detects recombinant rat Neuroligin 1/NLGN1 in direct ELISAs. Detects human and rat Neuroligin 1/NLGN1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.