Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat P-Selectin/CD62P Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF137
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF137 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF137 Supplier R&D Systems Supplier No. AF137
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 7 publications

P-Selectin/CD62P Polyclonal specifically detects P-Selectin/CD62P in Human, Rat samples. It is validated for Western Blot, Simple Western, Adhesion Blockade.

Specifications

Antigen P-Selectin/CD62P
Applications Western Blot, Adhesion Blockade
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 50 ug/mL, Adhesion Blockade
Gene Alias CD62P, CD62P antigen, FLJ45155, GMP140, GRMP, PADGEM, PADGEMantigen CD62), PSEL, selectin P (granule membrane protein 140kDa, antigen CD62), SELP
Gene Symbols SELP
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human P-Selectin/CD62P
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6403
Test Specificity Detects human and rat P-Selectin/CD62P in Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse P-Selectin, and less than 1% cross-reactivity with recombinant human (rh) E-Selectin and rhL-Selectin is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.