Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat PTK7/CCK4 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4499G
This item is not returnable.
View return policy
Description
PTK7/CCK4 Monoclonal specifically detects PTK7/CCK4 in Human, Rat samples. It is validated for Flow Cytometry.Specifications
PTK7/CCK4 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CCK4, CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5754 | |
Human, Rat | |
Purified |
Flow Cytometry | |
525222 | |
Flow Cytometry 5 uL/10^6 cells | |
Q13308 | |
PTK7 | |
E. coli-derived recombinant human PTK7/CCK4 Ala31-Ser199 Accession # Q13308 | |
100 Tests | |
Primary | |
Detects human and rat PTK7/CCK4 in Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction