Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat PTP pi/PTPRU Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7475
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7475 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7475 Supplier R&D Systems Supplier No. MAB7475
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTP pi/PTPRU Monoclonal specifically detects PTP pi/PTPRU in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen PTP pi/PTPRU
Applications Western Blot
Classification Monoclonal
Clone 764209
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q92729
Gene Alias FMI, GLEPP1, hPTP-J, pancreatic carcinoma phosphatase 2, PCP2, PCP-2, pi R-PTP-Psi, protein tyrosine phosphatase J, protein tyrosine phosphatase, receptor type, U, protein-tyrosine phosphatase J, protein-tyrosine phosphatase pi, PTP, PTP pi, PTP-J, PTP-PI, PTPPSI, PTPRO, PTP-RO, PTPRU, Receptor protein tyrosine phosphatase hPTP-J, R-PTP-PSI, R-PTP-U
Gene Symbols PTPRU
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PTP pi/PTPRU Glu19-Gln740 Accession # Q92729
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10076
Test Specificity Detects human and rat PTP pi/PTPRU in Western blots and recombinant human PTPRU in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) PTPRK, rhPTPRM, or rhPTPRT is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.