Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Human RBC Lysis Buffer PROMO
Click to view available options
:
500mL
Specifications
Specifications
Quantity | 500 mL |
Format | Liquid |
Product Type | Cell Lysis Reagent |
For Use With (Application) | Lysis of Human Red Blood Cells |
Content And Storage | 2° to 8°C |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction