Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human RBP4/Retinol-Binding Protein 4 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC3378T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3378T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC3378T100 Supplier R&D Systems Supplier No. IC3378T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Applications Flow Cytometry
Classification Monoclonal
Clone 393005
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P02753
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma
Gene Symbols RBP4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human RBP4/Retinol-Binding Protein 4 Glu19-Leu201 Accession # P02753
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5950
Test Specificity Detects human RBP4/Retinol-Binding Protein 4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 25% cross-reactivity with recombinant mouse RBP4 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.