Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RCOR1/CoREST Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6047
This item is not returnable.
View return policy
Description
RCOR1/CoREST Monoclonal specifically detects RCOR1/CoREST in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
RCOR1/CoREST | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q9UKL0 | |
RCOR1 | |
E. coli-derived recombinant human RCOR1/CoREST Ala394-Ser482 Accession # Q9UKL0 | |
100 μg | |
Primary | |
Detects human RCOR1/CoREST in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human RCOR3 is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
658526 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CoREST, KIAA0071CORESTRCORREST corepressor, Protein CoREST, RCOR1, REST corepressor 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
23186 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction