Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Reelin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Reelin |
---|---|
Clone | 940209 |
Dilution | Immunohistochemistry 5-15 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Description
Reelin Monoclonal specifically detects Reelin in Human samples. It is validated for Immunohistochemistry.Specifications
Reelin | |
Immunohistochemistry 5-15 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.21, EC 3.4.21.-, Reeler, reelin, RELN, RL, RLPRO1598 | |
RELN | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
940209 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P78509 | |
5649 | |
Chinese hamster ovary cell line CHO-derived recombinant human Reelin Ser1221-Gln2666 Accession # P78509 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Reelin in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title