Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Reg1A Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC4937U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4937U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC4937U100 Supplier R&D Systems Supplier No. IC4937U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Reg1A Monoclonal specifically detects Reg1A in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Reg1A
Applications Flow Cytometry
Classification Monoclonal
Clone 431202
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P05451
Gene Symbols REG1A
Host Species Rat
Immunogen E. coli-derived recombinant human Reg1A Gln23-Asn166 Accession # P05451
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human Reg1A in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Reg1B, rhReg4, recombinant rat (rr) Reg2, or rrReg3.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.