Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Reg1A/B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4937SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4937SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4937SP Supplier R&D Systems Supplier No. AF4937SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Reg1A Polyclonal specifically detects Reg1A in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Reg1A
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P05451
Gene Symbols REG1A
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Reg1A Gln23-Asn166 Accession # P05451
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5967
Test Specificity Detects human Reg1A in direct ELISAs. In direct ELISAs, approximately 80% cross-reactivity with recombinant human (rh) Reg1B is observed, approximately 20% cross-reactivity with recombinant mouse Reg1, and less than 5% cross-reactivity with rhReg3A and recombinant rat Reg1A is observed. 100% cross-reactivity with rhReg1B in Western blot is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.