Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Reg1A/B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB49371
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB49371 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB49371 Supplier R&D Systems Supplier No. MAB49371
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Reg1A Monoclonal specifically detects Reg1A in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Reg1A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 431211
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P05451
Gene Symbols REG1A
Host Species Rat
Immunogen E. coli-derived recombinant human Reg1A Gln23-Asn166 Accession # P05451
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human Reg1A in ELISAs. 100% cross-reactivity with recombinant human (rh) Reg1B in Western blot is observed. In direct ELISAs no cross-reactivity with rhReg3A, recombinant mouse Reg1, or recombinant rat Reg1A is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.