Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Reg1B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2090SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2090SP 25 μg
AF2090 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2090SP Supplier R&D Systems Supplier No. AF2090SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Reg1B Polyclonal specifically detects Reg1B in Human samples. It is validated for Western Blot.

Specifications

Antigen Reg1B
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P48304
Gene Alias Lithostathine 1 beta, lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2, PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2
Gene Symbols REG1B
Host Species Goat
Immunogen E. coli-derived recombinant human Reg1B Gln23-Asn166 Accession # P48304
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5968
Test Specificity Detects human Reg1B in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) Reg1 is observed, 5% cross-reactivity with rmReg2 and recombinant rat Reg1 is observed and less than 1% cross-reactivity with recombinant human Reg4 and rmReg3A is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.