Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Reg3A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5965
This item is not returnable.
View return policy
Description
Reg3A Monoclonal specifically detects Reg3A in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Reg3A | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q06141 | |
REG3A | |
E. coli-derived recombinant human Reg3A Glu27-Asp175 Accession # Q06141 | |
100 μg | |
Primary | |
Detects human Reg3A in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
512124 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hepatocarcinoma-intestine-pancreas, HIPpancreatitis-associated protein, Human proislet peptide, Lithostathine 3, pancreatic beta cell growth factor, Pancreatitis-associated protein 1, PAP homologous protein, PAP1FLJ18565, PAP2, PAPPAP-H, PBCGF, proliferation-inducing protein 34, proliferation-inducing protein 42, Reg III-alpha, REG3, REG3A, REG-3-alpha, regenerating islet-derived 3 alpha, regenerating islet-derived protein 3-alpha, Regenerating islet-derived protein III-alpha, REG-III | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5068 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction