Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Reg4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $667.00
Specifications
Antigen | Reg4 |
---|---|
Clone | 200214 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Reg4 Monoclonal specifically detects Reg4 in Human samples. It is validated for Western Blot.Specifications
Reg4 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9BYZ8 | |
83998 | |
E. coli-derived recombinant human Reg4 Asp23-Pro158 Accession # Q9BYZ8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Reg4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) Reg1, rmReg2, recombinant rat (rr) Reg3B, rmReg3A, rrReg3A, or rhReg1B. |
200214 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Gastrointestinal secretory protein, GISP, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELP, RELPREG-like protein | |
REG4 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title