Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Regulatory T Cell Cocktail Mouse anti-Human, Alexa Fluor 647, FITC, PE-Cy7, Clone: hIL-7R-M21, 2A3, SK3, BD

Catalog No. BDB560249 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560249 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560249 Supplier BD Biosciences Supplier No. 560249
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Human Regulatory T cell cocktail is a three color reagent used to identify the natural T regulatory cell (nTreg) population. An expression pattern of CD4+CD25int/brightCD127dim closely correlates to the expression of the transcription factor Forkhead box P3 (FoxP3). FoxP3 is considered a specific marker to T regulatory cells (Tregs). The SK3 antibody recognizes CD4, an antigen expressed primarily on helper/inducer T lymphocytes, although this antigen is also present in low density on monocytes. The 2A3 antibody recognizes CD25, an antigen also known as the human low-affinity interleukin-2 receptor (IL-2R). The CD25 antigen is present on a subset of peripheral blood lymphocytes. Antigen density increases on phytohemagglutinin (PHA)-, concanavalin A (Con A)-, and CD3-activated T lymphocytes; T lymphocytes from mixed lymphocyte cultures; and human T-lymphocyte leukemia virus (HTLV)-infected T-lymphocyte leukemia lines. hIL-7R-M21 reacts with the 60 - 90 kDa glycoprotein, CD127. CD127 is also known as the IL-7 receptor alpha (IL-7Rα). The receptor is a heterodimer composed of the CD127 and the common gamma chain, shared by other cytokine receptors (IL-2R, IL-4R, IL-9R, and IL-15R). CD127 is expressed on thymocytes, T- and B-cell progenitors, mature T cells, and some lymphoid and myeloid cells. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. The antibody was conjugated with PE-Cy7 under optimum conditions, and unconjugated antibody and free PE-Cy7 were removed. The antibody was conjugated with FITC under optimum conditions, and unreacted FITC was removed.

Cocktail components: FITC anti-Human CD4, PE-Cy7 anti-Human CD25, Alexa Fluor™ 647 anti-Human CD127
Clone: SK3, 2A3, hIL-7R-M21
IsoType: Isotype: Mouse IgG1, κ
Format: Isotype: FITC, PE-Cy™7, Alexa Fluor™ 647

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Regulatory T Cell Cocktail
Applications Flow Cytometry
Classification Monoclonal
Clone hIL-7R-M21, 2A3, SK3
Conjugate Alexa Fluor 647/FITC/PE-Cyanine7
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.