Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RelA/NFkB p65 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC50781S100UG
This item is not returnable.
View return policy
Description
RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| RelA/NFkB p65 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_068810 | |
| RELA | |
| E. coli-derived recombinant human RelA/NF kappa B p65 Met1-Thr254 Accession # NP_068810 | |
| RUO | |
| 5970 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 465003 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, p65RelA, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects human RelA/NF kappa B p65 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction