Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Relaxin-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$164.60 - $402.83
Specifications
| Antigen | Relaxin-1 |
|---|---|
| Clone | 361505 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB32571SP
![]() |
R&D Systems
MAB32571SP |
25 μg |
Each for $164.60
|
|
|||||
MAB32571
![]() |
R&D Systems
MAB32571 |
100 μg |
Each for $402.83
|
|
|||||
Description
Relaxin-1 Monoclonal specifically detects Relaxin-1 in Human samples. It is validated for Western Blot.Specifications
| Relaxin-1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human | |
| P04808 | |
| 6013 | |
| E. coli-derived recombinant human Relaxin-1 Lys26-Cys185 Accession # P04808 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects both the mature form and the proform of human Relaxin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Relaxin-2 or rhRelaxin-3 is observed. |
| 361505 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bA12D24.3.1, bA12D24.3.2, H1, preprorelaxin H1, prorelaxin H1, relaxin 1, relaxin 1 (H1), relaxin H1, Relaxin1, RLN1, RLXH1 | |
| RLN1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title