Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Relaxin-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB31071SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB31071SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB31071SP Supplier R&D Systems Supplier No. MAB31071SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Relaxin-3 Monoclonal specifically detects Relaxin-3 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Relaxin-3
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 332105
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 1-5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8WXF3
Gene Alias H3, INSL7, M3, relaxin 3, Relaxin3, RLN3, RXN3, ZINS4
Gene Symbols RLN3
Host Species Mouse
Immunogen E. coli-derived recombinant human Relaxin-3 Arg26-Cys142 Accession # Q8WXF3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117579
Test Specificity Recognizes the pro and mature forms of human Relaxin-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with the pro forms of recombinant human (rh) Relaxin-1 or rhRelaxin-2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.