Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Relaxin R2 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB102501S100UG
This item is not returnable.
View return policy
Description
Relaxin R2/LGR8 Monoclonal specifically detects Relaxin R2/LGR8 in Human samples. It is validated for Flow Cytometry.Specifications
Relaxin R2/LGR8 | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
G protein coupled receptor affecting testicular descent, Gpr106, GPR106G-protein coupled receptor 106, Great, GREATRelaxin family peptide receptor 2, INSL3R, leucine-rich repeat-containing G protein-coupled receptor 8, Leucine-rich repeat-containing G-protein coupled receptor 8, Lgr8, LGR8.1, LGR8G-protein coupled receptor affecting testicular descent, relaxin receptor 2, relaxin/insulin-like family peptide receptor 2, RelaxinR2, RLN2R, Rxfp2, RXFPR2 | |
Chinese hamster ovary cell line CHO-derived recombinant human Relaxin R2, Met1-Asn416, Accession # Q8WXD0 | |
100 μg | |
Primary | |
Detects human Relaxin R2 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
1013903 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8WXD0 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
122042 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction