Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RelB Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2698SP
This item is not returnable.
View return policy
Description
RelB Monoclonal specifically detects RelB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
RelB | |
Monoclonal | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q01201 | |
RELB | |
E. coli-derived recombinant human RelB Gln382-Thr579 Accession # Q01201 | |
25 μg | |
Primary | |
Detects human RelB. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
315206 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IREL, I-REL, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, v-rel reticuloendotheliosis viral oncogene homolog B | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5971 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction