Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human RELM beta Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $468.00
Specifications
Antigen | RELM beta |
---|---|
Clone | 250911 |
Concentration | LYOPH |
Dilution | ELISA |
Applications | ELISA |
Description
RELM beta Monoclonal specifically detects RELM beta in Human samples. It is validated for ELISA.Specifications
RELM beta | |
LYOPH | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9BQ08 | |
84666 | |
E. coli-derived recombinant human RELM beta Gly21-Thr111 Accession # Q9BQ08 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human RELM beta in direct ELISAs. Does not cross-react with recombinant mouse (rm) RELM alpha , rmRELM beta , recombinant rat RELM gamma , or rmResistin. Not recommended for natural lysates in Western blot. |
250911 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2 | |
RETNLB | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title